Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon, plasma standard solution, Specpure™ Si 10,000μg/mL

Catalog No. AA38723AD
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38723AD 50 mL
AA38723AE 100 mL
AA38723AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA38723AD Supplier Thermo Scientific Chemicals Supplier No. 038723AD
Add to Cart

Silicon plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00085311

Specifications

Physical Form Liquid
Solubility Miscible with molten alkali oxides, a mixture of nitric and hydrofluoric acids; Immiscible with water, nitric and hydrochloric acid
Quantity 50 mL
Grade Specpure
Chemical Name or Material Silicon plasma standard solution, Si 10000μg/mL
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.