Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon Standard, 1000mg/L in H2O/0.24 F (Assurance Grade), SPEX CertiPrep™
SDP

Catalog No. PLSI92T Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
30 mL
250 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PLSI92T 250 mL Poly Bottle
PLSI92M 30 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PLSI92T Supplier SPEX CertiPrep Supplier No. PLSI92T
Add to Cart
Add to Cart

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Silicon Standard
  • Made with ASTM Type I Water
  • 1,000 μg/mL concentration
  • 250 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Chemical Identifiers

CAS 16919-19-0 , 7732-18-5
Molecular Formula F6H8N2Si
Molecular Weight (g/mol) 178.153
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
PubChem CID 28145
IUPAC Name diazanium;hexafluorosilicon(2-)
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F

Specifications

Packaging Poly Bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Technique AA, ICP
Type Certified Reference Material
Quantity 250 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.H2O/tr. HF    1000μg/mL    PLSI9-2Y, -2T, -2X    10,000    PLSI9-3Y, -3XH2O    1000μg/mL    PLSI9A-2X    10,000    PLSI9A-3X
Chemical Name or Material Silicon Standard
Hazard Category Causes skin irritation, serious eye irritation. May cause respiratory irritation
DOTInformation Nonhazardous
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.