Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicone Fluid, 1000, Spectrum™ Chemical
SDP

Catalog No. 18606222 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
44 lb.
440 lb.
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-222 44 lb. Amber Glass Bottle
18-606-220 100 g Amber Glass Bottle
18-606-223 440 lb. Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-222 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SI11444LBBL
Add to Cart
Add to Cart

SI114, 63148-62-9

Spectrum™ Chemical Silicone Fluid, 1000

Chemical Identifiers

CAS 63148-62-9
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00132673
IUPAC Name Polydimethylsiloxane
SMILES C[Si](C)(-*)O-*

Specifications

CAS Max % 1
Quantity 44 lb.
Packaging Amber Glass Bottle
Viscosity 950-1050 cSt
Grade Reagent
Chemical Name or Material Silicone Fluid, 1000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.