Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silicone oil

Catalog No. 3783721L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-837-21L 1 L
37-837-2250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-837-21L Supplier Sigma Aldrich Supplier No. 3783721L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silicone oil is a liquid based siloxane that is part of the methyl silicone fluid system. It has a viscosity of 350 cSt with a refractive index of ∼ 1.402 and a dielectric strength of ∼ 14 kV/mm. It's surface tension tends to increase with an increase in the viscosity.Silicone oil can be used for a variety of applications such as: heat transferring medium in chemical and petrochemical industries, a dielectric coolant, a lubricant and antiflatulent agent, protective coatings for building materials, a cosmetic additive.

Specifications

Boiling Point >140°C/0.002, mmHg
CAS 63148-62-9
Quantity 1 L
Linear Formula [-Si(CH3)2 O-]n
MDL Number MFCD00132673
Refractive Index n20/D 1.403 (literature)
Density 0.968 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.