Learn More
World Precision Instrument Silicone Pad for use with ATC2000
SDP
Supplier:  World Precision Instrument 503573
Encompass_Preferred
ATC2000 is a low noise heating system for maintaining animal body temperature during experimental procedures. The DC heater is extremely quiet in terms of electromagnetic radiation. This is essential in electrophysiological recordings which are very sensitive to electromagnetic interference.
Catalog No. 50-253-9469
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.