Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Silicone Rubber Hand Protector
SDPSCI_ED

Catalog No. S27814 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S27814 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S27814 Supplier United Scientific™ Supplier No. HNDPTC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hand Protector constructed of silicone rubber with a specially textured grip for secure handling of hot objects such as beakers, flasks, and bottles.

  • Silicone rubber is capable of withstanding high temperatures.
  • Specially textured grip prevents containers from slipping.
  • Ideal for gripping hot objects such as beakers, flasks, and bottles.

Specifications

Accelerator Free No
Applications Laboratory
Coating Material Silicone
Color Red
Latex Free Yes
Material Silicone Rubber
Product Type Hand/Finger Protector
Size Universal
Style Full-hand Mitt
Temperature (English) Max. 350°F
Temperature (Metric) Max 176.6°C
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.