Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Silicone Sealing Mats for Deep Well Plates, 96 Round

Catalog No. 01183302 Shop All Simport Scientific Products
Change view
Click to view available options
Pre-slit:
No
Yes
Material:
Polyolefin, Rubber
Silicone
Pierceable:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pre-slit Material Pierceable
01-183-302 Yes Silicone Yes
01-183-296 No Polyolefin, Rubber No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-183-302 Supplier Simport™ Scientific Supplier No. T11063
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Impermeable sealing mats minimize evaporation in a wide range of temperatures in PCR and storage applications. These silicone mats are chemical-resistant, reusable, and provide a high degree of confidence.

  • Effectively seals deep well plates
  • Easy to open and close
  • Prevents the volatilization of a variety of solvents
  • Chemical and heat resistant
  • All pre-slit models allow the pipette tip to enter the hole without damage to the silicone cover
  • Resistant to DMSO and biological buffers
  • Free of RNase, DNase, Pyrogen, and DNA

Specifications

Color Natural
Solvent Or Alcohol Resistant Alcohol Resistant, Solvent Resistant
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
Pre-slit Yes
Certified RNase/DNase-free Yes
Pierceable Yes
Temperature Range (Metric) -80°C to +80°C
Well Shape Round
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.