Learn More
SOMNI Scientific™ Silicone Tubing
Silicone tubing provided custom cut per 12-inch increments to your specification. Silicone tubing is commonly used on anesthesia systems as it handles continuous bending/flexing and yet maintains flexibility over time
$13.01
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
04-777-181
![]() |
SOMNI Scientific™
TSB2009 |
Black |
Each for $13.01
|
|
|||||
04-777-187
![]() |
SOMNI Scientific™
TS2001 |
Opaque white |
Each for $13.01
|
|
|||||
Description
- Silicone tubing provided custom cut per 12 inch increments to your specification
- Silicone tubing is commonly used on anesthesia systems as it handles continuous bending and flexing
- Durable
- Maintains flexibility over time
- 3/16 in. ID
Specifications
3/16 in. | |
Sillicone |