Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Silodosin
GREENER_CHOICE

Catalog No. 666310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-631-0 10 mg
66-635-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 66-631-0 Supplier Tocris Bioscience™ Supplier No. 6663/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective α1A adrenoceptor antagonist (Ki = 0.036 nM)

Silodosin is a selective α1A adrenoceptor antagonist (Ki = 0.036 nM). Displays >55 fold selectivity for α1A over α1B and α1D. Inhibits norepinephrine-induced [Ca2+]I increase in α1A expressing CHO cells (IC50 = 0.32 nM). Inhibits norepinephrine-induced contraction of isolated human prostate tissue.

Specifications

Chemical Name or Material 2,3-Dihydro-1-(3-hydroxypropyl)-5-[(2R)-2-[[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl]amino]propyl]-1H-indole-7-carboxamide
CAS 160970-54-7
Quantity 10 mg
Molecular Formula C25H32F3N3O4
Formula Weight 495.53
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.