Learn More
Perkin Elmer US LLC SilQ+ MEINHARD Plus Nebulizer for Optima/Avio and NexION 300/350
Supplier:  Perkin Elmer US LLC N8122460
Encompass_Preferred
This MEINHARD nebulizer features a self aspiration rate of 0.5 mL/min with argon flow 1 L/min at 50 PSI (3.4 bar/345 kPa). Each nebulizer comes with the FitKit+ gas and sample quick connect fittings. The connections are made with high-purity PFA, minimizing the chance of contamination during backflow events. The gas connection fits snugly over the maria on the sidearm of the nebulizer.
Catalog No. 50-211-7208
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.