Learn More
Trajan Scientific SILTITE MICRO UNION FRRULE10PK
SDP
Supplier:  Trajan Scientific 073567
Encompass_Preferred
Trajan GC Capillary Column Connectors: Replacement Ferrules, For Use With: 0.1 to 0.25 mm I.D. First Column with 0.4 mm Holes and 0.32 mm I.D. Second Column with 0.5 mm Holes, Replacement SilTite u-Union Ferrules, Stainless Steel
Catalog No. 50-253-0194