Learn More
Sigma Organic Chemistry Silver | 25ML
Supplier: Sigma Organic Chemistry 80671425ML

Silver, 25ML
About This Item:
UNSPSC Code: 12352200
Storage: 2-8C
EINECS Number: 200-661-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.