Learn More
Medchemexpress LLC Silver, (diethylcarbamodithioato-κS,κS′)- | 1470-61-7 | MFCD00004929 | 99.2% | 256.14 | C5H10AgNS2 | 5 G

Supplier: Medchemexpress LLC HYW0346745G
Silver diethyldithiocarbamate is the silver salt of diethyldithiocarbamic acid, supplied as a solid organosilver reagent for biochemical and analytical applications. It is used as an analytical reagent and in biochemical assays that utilize silver-dithiocarbamate chemistry.
- Organosilver reagent for biochemical and analytical applications.
- High purity (99.15%) for reproducible results.
- Chemical formula C5H10AgNS2 and molecular weight 256.14.
- Available in small-scale packages starting at 5 g for laboratory use.
- Storage recommendations: 4°C, protect from light; in solution, store at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.