Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS Silver Disc Membrane

Catalog No. 09924069 Shop All Osmonics Products
Change view
Click to view available options
Pore Size:
1.2 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-924-069 1.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-924-069 Supplier GVS Supplier No. 1145345
Only null left
Add to Cart
Add to Cart
Request a Quote

Silver membranes are composed entirely of 99.97% pure metallic silver. They provide excellent chemical resistance and high temperature characteristics.

Properties: Silver membranes are composed entirely of 99.97% pure metallic silver. They provide excellent chemical resistance and high temperature characteristics. Orientation of the membrane can be important. There is a distinct difference in surface characteristics with one side appearing shinier than the other. Use the Shinier side upstream for scanning electron microscopy. For all other applications and analytical work use the duller side upstream.

Applications

  • Airborne asbestos fibers by X-Ray diffraction
  • Dissolved Organic Carbons (DOC)
  • Analysis of airborne silica in foundries, glass plants,
  • Quarries, mines, ceramic manufacturing
  • Coke oven emissions analysis
  • Carbon and carbon black
  • Coal tar pitch volatiles
  • Fly Ash - high temperature
  • Bromine and Chlorine analysis

Features and Benefits:

  • High temperature resistance
  • High chemical resistance
  • Hydrophilic
  • Economical - can be cleaned and reused
  • Autoclavable

Specifications

Autoclavable Autoclavable
Disposable Limited-use
Filter Type Membrane Filter
Format Disc
Material Silver
Pore Size 1.2 μm
Sterility Non-sterile
Type Disc Membrane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.