Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) behenate

Catalog No. AA4549422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4549422 100 g
AA4549406 5 g
AA4549414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4549422 Supplier Thermo Scientific Chemicals Supplier No. 4549422

CAS: 2489-05-6 | C22H43AgO2 | 447.45 g/mol

Silver(I) behenate is used as a calibration standard in X-ray spectrophotometer to an absolute X-ray energy scale. It is utilized for low-angle diffraction standard in X-ray crystallography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2489-05-6
MDL Number MFCD00059001

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 100 g
Molecular Formula C4H10ClNO2
Synonym n,n-dimethylglycine hydrochloride, dimethylglycine hydrochloride, glycine, n,n-dimethyl-, hydrochloride, n-methylsarcosine hydrochloride, n,n-dimethylglycine hcl, dimethylamino acetic acid hydrochloride, unii-yxk75eae92, yxk75eae92, n,n-dimethylaminoacetic acid hydrochloride, 2-dimethylamino acetic acid hydrochloride
Solubility Information Slightly soluble in water.
InChI Key FKASAVXZZLJTNX-UHFFFAOYSA-N
SMILES CN(C)CC(=O)O.Cl
IUPAC Name 2-(dimethylamino)acetic acid;hydrochloride
Molecular Weight (g/mol) 139.58
PubChem CID 75605
Formula Weight 447.44
Sensitivity Light sensitive
Chemical Name or Material Silver(I) behenate
Show More Show Less
EINECSNumber 219-641-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.