Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) behenate

Catalog No. AA4549414
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AA4549414

Supplier: Thermo Scientific Chemicals 04549414

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 2489-05-6 | C22H43AgO2 | 447.45 g/mol

Silver(I) behenate is used as a calibration standard in X-ray spectrophotometer to an absolute X-ray energy scale. It is utilized for low-angle diffraction standard in X-ray crystallography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2489-05-6
MDL Number MFCD00059001

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Molecular Formula C22H43AgO2
Synonym silver behenate, silver docosanoate, behenic acid silver salt, unii-n3hp26f3pb, n3hp26f3pb, silver hebenate, docosanoic acid, silver 1+ salt 1:1, docosanoic acid silver salt, docosanoic acid, silver salt, docosanoic acid, silver 1+ salt
Solubility Information Slightly soluble in water.
InChI Key AQRYNYUOKMNDDV-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)[O-].[Ag+]
IUPAC Name silver;docosanoate
Molecular Weight (g/mol) 447.45
PubChem CID 164971
Formula Weight 447.44
Sensitivity Light sensitive
Chemical Name or Material Silver(I) behenate
Show More Show Less
EINECSNumber 219-641-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.