Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver methanesulfonate

Catalog No. p-4824932
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1864109 10 g
AA1864118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1864109 Supplier Thermo Scientific Chemicals Supplier No. 01864109

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 2386-52-9 | CH3AgO3S | 202.96 g/mol

Silver methanesulfonate is used as a catalyst for heterocyclization reactions. Further, it is used in the synthesis of unsaturated ketones and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-52-9
Molecular Formula CH3AgO3S
Molecular Weight (g/mol) 202.96
MDL Number MFCD00064795
InChI Key MLKQJVFHEUORBO-UHFFFAOYSA-M
Synonym silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
PubChem CID 159658
IUPAC Name silver;methanesulfonate
SMILES CS(=O)(=O)[O-].[Ag+]

Specifications

Color White
Physical Form Powder
Quantity 10 g
Solubility Information Soluble in water,alcohol,ether and benzene. Slightly soluble in toluene,o-chlorotoluene and ethyl disulfide. Insoluble in hexane and methylcyclopentane.
Formula Weight 202.97
Odor Odorless
Sensitivity Light sensitive
Appearance White powder
Chemical Name or Material Silver methanesulfonate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9, Packing Group: III
EINECSNumber 219-199-2
RTECSNumber PB2685000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.