Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver molybdenum oxide, 99%

Catalog No. p-7050889
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3965001 0.25 g
AA3965003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3965001 Supplier Thermo Scientific Chemicals Supplier No. 03965001

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 13765-74-7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-74-7
MDL Number MFCD00053384
Synonym Silver(I) molybdate

Specifications

Melting Point 483°C
Color Beige
Physical Form Powder
Quantity 0.25 g
Formula Weight 375.68
Percent Purity 99%
Odor Odorless
Sensitivity Light sensitive
Density 6.18 g/mL
Chemical Name or Material Silver molybdenum oxide
EINECSNumber 237-374-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.