Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.100 N (N/10), Ricca Chemical
SDP

Catalog No. R70000004C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Amber Poly Bottle
Black Poly Bottle
Cubitainer
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R70000004C 4 L Amber Glass Bottle
7000-4 120 mL Amber Poly Bottle
7000-16 500 mL Amber Poly Bottle
R70000001C 1 L Amber Glass Bottle
7000-32 1 L Amber Poly Bottle
7000-1 4 L Black Poly Bottle
R70000004F 4 L Cubitainer
7000-25 10 L Cubitainer
7000-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. R70000004C Supplier Ricca Chemical Company Supplier No. 70001G
Add to Cart
Add to Cart

Silver Nitrate Titrants

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
Boiling Point 100°C
CAS Min % 98.32, 1.68
Melting Point 0°C
CAS Max % 98.32, 1.68
Density 1.0145
Solubility Information Miscible in water
Grade Laboratory
Packaging Amber Glass Bottle
pH ∼6
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Silver Nitrate, 0.100 Normal (N/10)
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.