Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.250 Normal Standardized,, Reagents

Catalog No. 01333587
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-587 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-587 Supplier Reagents Holdings Llc Supplier No. CS1509004C
Add to Cart
Add to Cart

Silver Nitrate, 0.250 Normal Standardized, NIST Traceable, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Silver Nitrate, 0.250 Normal Standardized, NIST Traceable
Boiling Point 100°C
CAS 7732-18-5, 7761-88-8
Color Colorless
Physical Form Liquid
Quantity 4 L
Molecular Formula AgNO3/H2O
Synonym Nitric Acid silver salt
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0328
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.