Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, Crystal, USP, 99.8 -100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606238 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
250 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-238 500 g Amber Glass Bottle
18-606-237 125 g Amber Glass Bottle
18-606-239 250 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-238 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SI115500GM
Add to Cart
Add to Cart
Request a Quote

SI115, 7761-88-8, AgNO3

Spectrum™ Chemical Silver Nitrate, Crystal, USP is used as an antibiotic for indolent wounds, corn and callus removal and as a Gonorrheal ophthalmia neonatorum prophylaxis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99.8 to 100.5%
Grade USP
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.