Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate Titrant, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88767 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88767 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88767 Supplier Aqua Phoenix Scientific Supplier No. SN3422B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Boiling Point 100°C to 102°C
Color Colorless
CAS 7761-88-8, 7732-18-5
Quantity 60 mL
Molecular Formula AgNO3
Linear Formula AgNO3
Synonym Silver nitrate
Solubility Information Miscible in Water
PubChem CID 24470
Odor Odorless
Appearance Colorless Liquid
Density 1.02 g/cm³ (at 20°C)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.