Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Standard, 10,000μg/mL in water w/Dilute Nitric Acid, Ultra Scientific

Catalog No. USICP147
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-ICP-147 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-ICP-147 Supplier Agilent Technologies Supplier No. ICP147
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7440-22-4 , 7697-37-2
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Physical Form Liquid
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 10, 000μg/mL
Chemical Name or Material Silver Standard, 10, 000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.