Learn More
Silver sulfate, Premion™, 99.999% (metals basis)
Description
Silver(I) sulfate is employed in silver plating, and in electrolysis. It is a useful non-staining substitute to silver nitrate. Silver sulfate/iodine is an excellent iodination reagent for alkyl, alkoxybenzenes, aromatic amines and uridines. It is used in batteries; its pharmaceutical composition is used as an antimicrobial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1085°C (decomposition) |
| Melting Point | 657°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.999% (metals basis) |
| Solubility Information | Partially soluble in water. Water solubility increases with temperature. Soluble in nitric acid,ammonia and conc. sulfuric acid. |
| Formula Weight | 311.8 |
| Odor | Odorless |
| Sensitivity | Light sensitive |
| Show More |
Safety and Handling
| Hazard Category | H318 |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| DOTInformation | DOT Class: 9, Packaging Group: III |
| EINECSNumber | 233-653-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.