Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver sulfate, Premion™, 99.999% (metals basis)

Catalog No. AA1211222
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol

Silver(I) sulfate is employed in silver plating, and in electrolysis. It is a useful non-staining substitute to silver nitrate. Silver sulfate/iodine is an excellent iodination reagent for alkyl, alkoxybenzenes, aromatic amines and uridines. It is used in batteries; its pharmaceutical composition is used as an antimicrobial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
MDL Number MFCD00003407
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Boiling Point 1085°C (decomposition)
Melting Point 657°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.999% (metals basis)
Solubility Information Partially soluble in water. Water solubility increases with temperature. Soluble in nitric acid,ammonia and conc. sulfuric acid.
Formula Weight 311.8
Odor Odorless
Sensitivity Light sensitive
Appearance White powder
Density 5.45 g/cm3 at 20°C
Chemical Name or Material Silver sulfate
Show More Show Less
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 233-653-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.