Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silvex™ (2,4,5-TP) (For EPA 500, 8000/; Appendix IX Methods)

Catalog No. USHB1101
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-HB-110-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-HB-110-1 Supplier Agilent Technologies Supplier No. HB1101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Silvex™ at 100μg/mL in methanol

Chemical Identifiers

CAS 93-72-1
Molecular Formula C9H7Cl3O3
Molecular Weight (g/mol) 269.50
MDL Number MFCD00002646
InChI Key ZLSWBLPERHFHIS-UHFFFAOYNA-N
PubChem CID 7158
ChEBI CHEBI:34758
IUPAC Name 2-(2,4,5-trichlorophenoxy)propanoic acid
SMILES CC(OC1=CC(Cl)=C(Cl)C=C1Cl)C(O)=O

Specifications

Packaging Ampule
Quantity 1 x 1 mL
Chemical Name or Material Silvex™ (2, 4, 5-TP)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.