Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Silylation Grade Solvents

Catalog No. pi20672
Encompass
Change view
Click to view available options
Standard Components:
Acetonitrile
Dimethylformamide (DMF)
Dimethylsulfoxide (DMSO)
Pyridine
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Components
PI20672 Dimethylformamide (DMF)
PI20062 Acetonitrile
PI20684 Dimethylsulfoxide (DMSO)
PI27530 Pyridine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI20672 Supplier Thermo Scientific™ Supplier No. TS20672
Only null left
Add to Cart
Add to Cart

Use specially manufactured and packaged Thermo Scientific™ Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions.

Use specially manufactured and packaged Thermo Scientific™ Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions. They are purified, dried, and packaged under nitrogen in convenient Thermo Scientific™ Hypo-Vial™ Sample Storage Vials, and are supplied with elastomer septa to enable immediate access to the sample without exposure to moisture and oxygen. Use polar solvents (acetonitrile, dimethylformamide, dimethylsulfoxide, pyridine, tetrahyrdrofuran) to facilitate reactions; nonpolar organic solvents may be used but will not accelerate the rate of reaction.

Recommended Use

Avoid water or alcohol, as TMS reagents react with active hydrogen.

Avoid enolizable ketones.

Use dimethylformamide for steroids and other large molecules.

Use dimethylsulfoxide to prepare TMS derivatives of tertiary alcohols and some compounds with low solubility in other silylation solvents.

Pyridine is an excellent solvent and reaction medium for MS reactions and is an HCI acceptor in reactions involving organochlorosilanes.

Other commonly used solvents include tetrahydrofuran and acetonitrile.

TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Hazards Flammable, Harmful
Item Description Dimethylformamide (DMF); 50 mL
Standard Components Dimethylformamide (DMF)
Standard Concentration 98 to 100%
Storage 4°C

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.