Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simazine, Restek
SDP

Catalog No. 06719863 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-863 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-863 Supplier Restek Corporation Supplier No. 32236
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 122-34-9
Molecular Formula C7H12ClN5
Molecular Weight (g/mol) 201.658
InChI Key ODCWYMIRDDJXKW-UHFFFAOYSA-N
PubChem CID 5216
ChEBI CHEBI:27496
IUPAC Name 6-chloro-2-N,4-N-diethyl-1,3,5-triazine-2,4-diamine
SMILES CCNC1=NC(=NC(=N1)Cl)NCC

Specifications

Quantity 1 mL
Chemical Name or Material Simazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.