Learn More
United Scientific™ Simple Anemometer
SDPSCI_ED
Catalog No. S139519 Shop All United Scientific Supplies, Inc Products
This item is not returnable. View return policy
Catalog No. S139519 Supplier United Scientific™ Supplier No. ANMM01
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Simple Anemometer is a useful tool for determining wind speed.
  • This simple tool measures wind speed and direction.
  • The wind speed range is 0 to 17 meters per second (±10%).
  • The direction indicator is divided into 16 compass points.
  • A built-in compass is provided.
Specifications
Includes Built-in Compass
Product Type Anemometer
Warning: Cancer and reproductive harm
Prop 65