Description
Includes
Half-meter truss with hooks on both sides, V-shaped clamp and instructions
Basic model provides an interesting introduction to simple bridge building and explores compressions, tensions and force and equilibrium.
Order Info
Stand, masses and spring scale sold separately.
Specifications
Specifications
| Product Type | Simple Truss |
| Material | Wood |
| For Use With (Application) | Demonstrate force and equilibrium |
| Includes | Half meter truss with hooks on both sides, V-shaped clamp and instructions |