Learn More
Eisco™ Simple Form Truss with Screw Clamp
SCI_ED
Catalog No. S27665 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S27665 1 Ea.
1 options
Catalog No. S27665 Supplier Eisco™ Supplier No. PH0344SFT
Add to Cart
Add to Cart
This Simple Form Truss with Screw Clamp at one end allows for the measurement of force, as well as the observation of the effect of force. It's a great tool for group projects and individual study exploring structural engineering, static equilibrium, levers and torque.
  • Meter stick features measurements from 1 to 50 cm on both sides
  • Required accessories are all sold separately: rod/stand, spring scale, and masses
  • Includes 8 Hooks (4 on each side)
Specifications
Product Type Simple Form Truss
Description Clamp at one end
For Use With (Application) Measuring and Observing The Effects of Force
For Use With (Equipment) Rod/Stand, Spring Scale, and Masses (Sold Separately)
Includes Screw Clamp
Quantity 1 Ea.
Length (English) 22 in.