Learn More
R&D Systems Simple Plex Cartridge Kit, automated ELISA, single analyte, human IFN-gamma, volume 10 μL, sensitivity 0.05 pg/mL, assay range 0.2 - 4000 pg/mL, assay length 90 min
SDP
Catalog No. NC3628003 Shop All Novus Biologicals Products
Encompass
Simple Plex Cartridge Kit, automated ELISA, single analyte, human IFN-gamma, volume 10 μL, sensitivity 0.05 pg/mL, assay range 0.2 - 4000 pg/mL, assay length 90 min
Catalog No. NC3628003 Supplier R&D Systems Supplier No. SPCKBCS002697
Add to Cart
Add to Cart
Simple Plex Cartridge Kit, automated ELISA, single analyte, human IFN-gamma, volume 10 μL, sensitivity 0.05 pg/mL, assay range 0.2 - 4000 pg/mL, assay length 90 min