Learn More
R&D Systems Simple Plex Control for Mouse/Rat Angiopoietin-2
SDP
Catalog No. NC3654250 Shop All Novus Biologicals Products
Encompass
INTENDED USE AND DESCRIPTION: For use a quantitative control for the determination of mouse/rat Angiopoietin-2 concentrations in biological fluids. Controls are supplied lyophilized. Please refer to the lot specific certificate of analysis and Simple Plex Lyophilized Quality Control product insert for additional information.Angiopoietins, Ang-1, Ang-2, and Ang-3 (mouse)/Ang-4 (human), are natural agonists or antagonists of the Tie-2 receptor tyrosine kinase and are important modulators of angiogenesis. Two domains characterize the angiopoietin family of proteins: an N-terminal coiled-coil domain that mediates homo-oligomerization, and a C-terminal fibrinogen-like domain that binds Tie-2.
Catalog No. NC3654250 Supplier R&D Systems Supplier No. 896173
Customers require a valid business account associated to their profile to access pricing and ordering options.
INTENDED USE AND DESCRIPTION: For use a quantitative control for the determination of mouse/rat Angiopoietin-2 concentrations in biological fluids. Controls are supplied lyophilized. Please refer to the lot specific certificate of analysis and Simple Plex Lyophilized Quality Control product insert for additional information.Angiopoietins, Ang-1, Ang-2, and Ang-3 (mouse)/Ang-4 (human), are natural agonists or antagonists of the Tie-2 receptor tyrosine kinase and are important modulators of angiogenesis. Two domains characterize the angiopoietin family of proteins: an N-terminal coiled-coil domain that mediates homo-oligomerization, and a C-terminal fibrinogen-like domain that binds Tie-2.