Learn More
Specifications
Specifications
| Quantity | 1 Ea. |
| Species | Non-Human Primate |
| Binding Target | GFAP |
| Gene ID (Entrez) | 2670 |
| Synonym | ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein |
| Content And Storage | Store lyophilized controls at 2°C to 8°C. Lyophilized controls are stable for up to 12 months from date of receipt. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
