Learn More
R&D Systems SIMPLE PLEX DEMO KIT COLD
SDP
Catalog No. NC3588990
Encompass
Simple Plex demo kit includes cartridges (2) (SPCKAPS-011808: CCL2/MCP-1, IL-6 3rd Gen, TNF-alpha 2nd Gen,and VEGF-A), controls, and diluents. Store at 4c.
Catalog No. NC3588990 Supplier R&D Systems Supplier No. 600102 COLD
Customers require a valid business account associated to their profile to access pricing and ordering options.
Simple Plex demo kit includes cartridges (2) (SPCKAPS-011808: CCL2/MCP-1, IL-6 3rd Gen, TNF-alpha 2nd Gen,and VEGF-A), controls, and diluents. Store at 4c.