Learn More
Specifications
Specifications
| Content And Storage | Store lyophilized controls at 2-8 °C. Lyophilized controls are stable for up to 12 months from date of receipt. Reconstituted controls are stable for up to 6 months from date of preparation when stored at ≤ -70 °C. Storage at -20° C is not recommended. Individual frozen aliquots of low and high controls are designed as single-use entities. Thaw directly before use and discard any remaining volume. |
| Detectable Analytes | CD23/Fc epsilon RII |
| Sample Type | Biological Fluids |
| Technology | Control Material |
| Gene ID (Entrez) | 2208 |
| Quantity | 1 Each |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
