Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Simple Plex Discovery Control for Human CG beta
SDP

Catalog No. 897214 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-721-4 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-721-4 Supplier R&D Systems™ Supplier No. 897214

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Control for use with Human CG beta Simple Plex Assay Cartridge Kit

Specifications

Content And Storage Store lyophilized controls at 2-8 °C. Lyophilized controls are stable for up to 12 months from date of receipt. Reconstituted controls are stable for up to 6 months from date of preparation when stored at ≤ -70 °C. Storage at -20° C is not recommended. Individual frozen aliquots of low and high controls are designed as single-use entities. Thaw directly before use and discard any remaining volume.
Detectable Analytes CG beta
Sample Type Biological Fluids
Technology Control Material
Gene ID (Entrez) 1082
Quantity 1 Each
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.