Learn More
R&D Systems Simple Plex Discovery Human Uteroglobin Assay, Automated sandwich ELISA, Microfluidic cartridge, Sample volume 10 μL, Assay length 90 min
SDP
Catalog No. NC3725588 Shop All Novus Biologicals Products
Encompass
Simple Plex Discovery Human Uteroglobin Assay, Automated sandwich ELISA, Microfluidic cartridge, Sample volume 10 μL, Assay length 90 min
Catalog No. NC3725588 Supplier R&D Systems Supplier No. SPCKBPS012701
Customers require a valid business account associated to their profile to access pricing and ordering options.
Simple Plex Discovery Human Uteroglobin Assay, Automated sandwich ELISA, Microfluidic cartridge, Sample volume 10 μL, Assay length 90 min