Learn More
R&D Systems™ Simple Plex Non-Human Primate Neurogranin Cartridge
Shop All Novus Biologicals Products

Specifications
Specifications
| Quantity | 32 Samples |
| Species | Non-Human Primate |
| Binding Target | Neurogranin |
| Assay Sensitivity | 4.23 pg/mL |
| Assay Range | 16.40 to 4000 pg/mL |
| Gene ID (Entrez) | 4900 |
| Product Line | ProteinSimple |
| Sample Type | Cell Culture Supernates, Serum, EDTA (Plasma), Heparin (Plasma), Cerebrospinal Fluid |
| Synonym | calmodulin-binding protein, hng, neurogranin, neurogranin (protein kinase C substrate, RC3), Ng, NRGN, protein kinase C substrate, RC3, RC3ng |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.