Learn More
Specifications
Specifications
| Quantity | 32 Samples |
| Species | Non-Human Primate |
| Binding Target | TREM2 |
| Assay Sensitivity | 1.78 pg/mL |
| Assay Range | 9.96 to 15200 pg/mL |
| Gene ID (Entrez) | 54209 |
| Product Line | ProteinSimple |
| Sample Type | Cell Culture Supernates, Serum, EDTA (Plasma), Heparin (Plasma), Cerebrospinal Fluid |
| Synonym | PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
