Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Simple Resonance Tube Set
SCI_ED

Catalog No. S12968 Shop All Eisco Products
Change view
Click to view available options
11.4L x 2.4W x 2.4 in. H
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S12968 11.4L x 2.4W x 2.4 in. H
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12968 Supplier Eisco™ Supplier No. PH0715
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Demonstrates the principles of resonance without requiring a water column or other apparatus.

  • Tuning fork with 512 Hz used as sound source
  • Inner tube slides inside outer tube
  • Tubes adjust from 25 to 38cm with tuning range of 452 to 585 Hz

Specifications

Product Type Simple Resonance Tube Set
For Use With (Application) Demonstrates the principles of resonance
Dimensions (L x W x H) 11.4 x 2.4 x 2.4 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.