Learn More
MilliporeSigma™ Simplicon™ Reprogramming RNA (OKSG)
Catalog No. SCR549 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SCR549 1 Kit
1 options
Catalog No. SCR549 Supplier MilliporeSigma™ Supplier No. SCR549
Add to Cart
Add to Cart
For use as transfection reagent.
  • Safe, efficient
  • Stem cell type: Induced pluripotent stem cells
  • Generates integration free, virus-free human iPS cell
  • Single transfection step of self-replicating synthetic RNA for Oct4, Klf4, Sox2, Glis1
  • Integration free, footprint free iPS cells
  • No risk of genomic integration
  • Safe, virus free, synthetic polycistronic RNA replicon (all four reprogramming factors in 1 RNA str)
  • Only 1 day transfection required
  • RNA replicon is able to self-replicate, elliminating need for laborious daily transfection of multiple individual mRNAs over 14 day period
  • Efficient rapid reprogramming
  • No screening required to ensure viral remnants are not present
  • Controlled elimination of synthetic RNA replicon by removal of B18R protein
  • Validated for reprogramming in feeder-based feeder-free culture conditions
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Content And Storage >-70°C