Learn More
MilliporeSigma™ Simplicon™ RNA Reprogramming Kit (OKSG)
Catalog No. SCR550 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SCR550 1 Kit
1 options
Catalog No. SCR550 Supplier MilliporeSigma™ Supplier No. SCR550
Add to Cart
Add to Cart
Includes
VEE-OKS-iG RNA, B18R RNA
For use as transfection reagent.
  • Safe efficient method
  • Generates integration free, virus-free human IPS cell
  • Single transfection step of self-replicating synthetic RNA for Oct4, Klf4, Sox2 Glis1
  • Integration-free, footprint-free iPS cells
  • No risk of genomic integration
  • Safe, virus free, synthetic polycistronic RNA replicon (all four reprogramming factors in 1 RNA str)
  • Only 1 day transfection required
  • Efficient rapid reprogramming
  • No screening required to ensure viral remnants are not present
  • Controlled elimination of synthetic RNA replicon by removal of B18R protein
  • Validated for reprogramming in feeder based feeder free culture conditions
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Content And Storage >-70°C