Description
- Flexible, polypropylene plates can be combined on thermal cycler block
- Thin-wall, 0.2mL wells with nonbinding inner surface
- Sealable with cap strips, sealing films, foils or mats
- Autoclavable
Certifications
Certified RNase-, DNase-, and DNA-free
Specifications
Specifications
| Format | Standard |
| Array | 6 x 8 |
| Color | Green |
| Material | Polypropylene |
| No. of Wells | 48 |