Learn More
Simport™ Scientific EasyDip™ Stainless-Steel Holder
Catalog No. 22038980 Shop All Simport Scientific Products
Change view
Click to view available options
Product Type:
Easydip Holder
Catalog No. Product Type
22-038-980 Easydip Holder
1 options
This item is not returnable. View return policy
Catalog No. 22-038-980 Supplier Simport™ Scientific Supplier No. M906
Add to Cart
Add to Cart
This item is not returnable. View return policy
The EasyDip™ Stainless-Steel Holder holds up to six EasyDip™ staining jars.
Sturdy holder, for use with the EasyDip™ Slide Staining System (Cat. Nos. 22-038-489 through -495).
Dimensions: 16 3/4 x 4 x 1 1/2 in. (42.5 x 10.2 x 3.8cm)
Specifications
Product Type Easydip Holder
For Use With (Equipment) 6 EasyDip™ slide staining systems