Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes
Catalog No. 22040517 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Peach
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-040-517 Aqua
22-040-519 Peach
22-040-521 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-040-517 Supplier Simport™ Scientific Supplier No. M48012
Add to Cart
Add to Cart
M480 Disposable Embedding Cassettes for holding and identifying tissue samples in processing, embedding and sectioning procedures during in vitro diagnostics.
  • Made of acetal polymer and resistant to chemical action of histological solvents
  • Fit securely in microtome chuck adapters
  • Accepts standard metal lids that keep specimens safe during processing
  • Slanted writing surface for easy sample identification
  • Available in three colors
Certifications
CE marked
Specifications
Color Aqua
Material Acetal Polymer
Product Type Embedding Cassette