Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes
Catalog No. 22040521 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Peach
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-040-521 White
22-040-517 Aqua
22-040-519 Peach
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-040-521 Supplier Simport™ Scientific Supplier No. M4802
Add to Cart
Add to Cart
This item is not returnable. View return policy
M480 Disposable Embedding Cassettes for holding and identifying tissue samples in processing, embedding and sectioning procedures during in vitro diagnostics.
  • Made of acetal polymer and resistant to chemical action of histological solvents
  • Fit securely in microtome chuck adapters
  • Accepts standard metal lids that keep specimens safe during processing
  • Slanted writing surface for easy sample identification
  • Available in three colors
Certifications
CE marked
Specifications
Color White
Material Acetal polymer
Product Type Embedding Cassette