Learn More
Simport™ Scientific CoreDish™ 12-Compartment General Purpose Biopsy Container
Shop All Simport Scientific ProductsDescription
- Reduces risk of confusion and protects from contamination
- Polystyrene made
- Screw on lid incorporates an O-ring
- Leakproof
- Compartments numbered from 1 to 12
- For general purpose
Specifications
Specifications
| Product Type | Biopsy Container |
| Material | Polystyrene |
| Closure Type | O-Ring Cap |
| For Use With (Application) | General Purpose Biopsy |