Learn More
Simport™ Scientific CoreDish™ 12-Compartment Lower GI Tract Biopsy Container
Shop All Simport Scientific ProductsDescription
- Safe and easy transport of specimens from collection to analysis
- O-ring lid
- Leakproof seal
- Twelve compartments
- An area for patient information
- Ten labeled compartments: Proximal Flexure Colon, Hepatic Flexure Colon, Distal Transverse Colon, Ascending Colon, Splenic flexure Colon, Cecum, Decending Colon, Terminal Ileum, Rectum, Sigmoid Colon
- For lower GI tract
Specifications
Specifications
| Product Type | Biopsy Container |
| Closure Type | O-Ring Cap |
| For Use With (Application) | Lower GI Tract Biopsy |
| Label | 10 |