Learn More
Simport™ Scientific CoreDish™ 8-Compartment Upper GI Tract Biopsy Container
Catalog No. 22045685 Shop All Simport Scientific Products
Change view
Click to view available options
Label:
7
Catalog No. Label
22-045-685 7
1 options
This item is not returnable. View return policy
Catalog No. 22-045-685 Supplier Simport™ Scientific Supplier No. M971D8UGI
Add to Cart
Add to Cart
This item is not returnable. View return policy
The CoreDish™ 8-Compartment Upper GI Tract Biopsy Container offers many configurations in order to hold different biopsies of the breast, prostate, upper GI tract, and lower GI tract. A label allows essential information to be written such as patient I.D., doctor, date, and time.
  • For safe and easy transport of the specimens from collection to analysis
  • Polystyrene made
  • O-ring lid for leakproof protection
  • An area for patient information on label
  • Seven labeled compartments: Gastric Card, Gastric Body, GE Junction, Gastric ATR, Distal Esophage, Pylorus, Duodeum
  • For upper GI tract
Specifications
Product Type Biopsy Container
Material Polystyrene
Closure Type O-Ring Cap
For Use With (Application) Upper GI Tract
Label 7